Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK2 Antibody (OTI2D9), PE, Novus Biologicals™
SDP

Catalog No. NBP270392PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270392PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270392PE Supplier Novus Biologicals Supplier No. NBP270392PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDK2 Monoclonal specifically detects CDK2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen CDK2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2D9
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2)
Gene Symbols CDK2
Host Species Mouse
Immunogen Full length recombinant protein of human CDK2 (NP_001789) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1017.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.