Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK20 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB319997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB319997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB319997 Supplier Novus Biologicals Supplier No. NBP298621PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDK20 Polyclonal antibody specifically detects CDK20 in Human samples. It is validated for Western Blot

Specifications

Antigen CDK20
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias CAK-kinase p42, CCRKEC 2.7.11.22, CDCHP42, CDK-activating kinase p42, cell cycle related kinase, Cell cycle-related kinase, Cell division protein kinase 20, cyclin-dependent kinase 20, Cyclin-dependent protein kinase H, Cyclin-kinase-activating kinase p42, EC 2.7.11, p42, PNQALRE
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CDK20 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23552
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.