Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK4 Antibody (6D10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP242642A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP242642A30 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP242642A30 Supplier Novus Biologicals Supplier No. NBP242642AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDK4 Monoclonal specifically detects CDK4 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CDK4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6D10
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4
Gene Symbols CDK4
Host Species Mouse
Immunogen Recombinant human CDK4 (1-303aa) purified from E. coli
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 1019
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.