Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK4 Antibody (DCS-31), Novus Biologicals™
SDP

Catalog No. NB1206315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206315 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206315 Supplier Novus Biologicals Supplier No. NB1206315
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDK4 Monoclonal specifically detects CDK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray.

Specifications

Antigen CDK4
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Microarray
Classification Monoclonal
Clone DCS-31
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Microarray
Gene Accession No. P11802
Gene Alias Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4
Gene Symbols CDK4
Host Species Mouse
Immunogen Recombinant human Cdk4 protein
Molecular Weight of Antigen 33 kDa
Purification Method Unpurified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1019
Test Specificity Reacts specifically with Cdk4. Does not recognize other Cdk types.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.