Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK4 Rabbit anti-Human, Clone: SD205-1, Novus Biologicals™
SDP

Catalog No. NBP267751 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267751 Supplier Novus Biologicals Supplier No. NBP267751

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

CDK4 Monoclonal specifically detects CDK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen CDK4
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SD205-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4
Gene Symbols CDK4
Host Species Rabbit
Immunogen Recombinant protein within mouse CDK4 aa 180-303. (SwissProt: P11802 Human; SwissProt: P30285 Mouse; SwissProt: P35426 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1019
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.