Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk5 Antibody (OTI2H6), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N270396APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270396APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270396APC7 Supplier Novus Biologicals Supplier No. NBP270396APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdk5 Monoclonal specifically detects Cdk5 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.

Specifications

Antigen Cdk5
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2H6
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit
Gene Symbols CDK5
Host Species Mouse
Immunogen Full length recombinant protein of human Cdk5 (NP_004926) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Innate Immunity, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1020.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.