Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk5 Rabbit anti-Human, Mouse, Rat, Clone: 7C8T10, Novus Biologicals™
SDP

Catalog No. NB166336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166336 20 μg
NB166335 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166336 Supplier Novus Biologicals Supplier No. NBP31562020UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cdk5 Monoclonal antibody specifically detects Cdk5 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Cdk5
Applications Western Blot
Classification Monoclonal
Clone 7C8T10
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 193-292 of human Cdk5 (Q00535).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1020
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.