Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdk5 Rabbit anti-Human, Mouse, Rat, Clone: 7C8T10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Cdk5 |
---|---|
Clone | 7C8T10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Cdk5 Monoclonal antibody specifically detects Cdk5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Cdk5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Innate Immunity | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1020 | |
IgG | |
Affinity purified |
7C8T10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit | |
A synthetic peptide corresponding to a sequence within amino acids 193-292 of human Cdk5 (Q00535). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title