Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk6 Antibody (8G3), Novus Biologicals™
SDP

Catalog No. NB297651 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297651 100 μL
NB297650 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297651 Supplier Novus Biologicals Supplier No. NBP326663100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cdk6 Monoclonal antibody specifically detects Cdk6 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Cdk6
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 8G3
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Cell division protein kinase 6, cyclin-dependent kinase 6, EC 2.7.11, EC 2.7.11.22, PLSTIREMGC59692, Serine/threonine-protein kinase PLSTIRE
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Cdk6 [UniProt Q00534]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1021
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.