Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdk6 Antibody (8G3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Cdk6 |
---|---|
Clone | 8G3 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
Cdk6 Monoclonal antibody specifically detects Cdk6 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
Cdk6 | |
Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cell division protein kinase 6, cyclin-dependent kinase 6, EC 2.7.11, EC 2.7.11.22, PLSTIREMGC59692, Serine/threonine-protein kinase PLSTIRE | |
A synthesized peptide derived from Human Cdk6 [UniProt Q00534] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
8G3 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1021 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title