Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk7 Rabbit anti-Human, Clone: JJ203-01, Novus Biologicals™
SDP

Catalog No. NBP267850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267850 Supplier Novus Biologicals Supplier No. NBP267850

Rabbit Monoclonal Antibody

Cdk7 Monoclonal specifically detects Cdk7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Cdk7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JJ203-01
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias CAK1Xenopus laevis, cdk-activating kinase), CDK-activating kinase, Cell division protein kinase 7, cyclin-dependent kinase 7, cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, EC 2.7.11.24, p39 Mo15, serine/threonine kinase stk1,39 kDa protein kinase, serine/threonine protein kinase 1, serine/threonine protein kinase MO15, TFIIH basal transcription factor complex kinase subunit
Gene Symbols CDK7
Host Species Rabbit
Immunogen Recombinant protein within Human Cdk7 aa 151-346/ 346. (SwissProt: P50613 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1022
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.