Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK8 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
Supplier: Active Motif 61481
Description
A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34/Cdc 2, and a related protein Cdk 2, function late in the cycle while Cdk 4 and Cdk 6 are critically involved in G1 to S progression. The p54/Cdk 8 interacts exclusively with Cyclin C. The Cdk 8 Cyclin C complex activates the kinase. Because of analogy with yeast Cdk 8, it has been proposed that the human Cdk 8 Cyclin C may interact with RNA polymerase, thus influencing the transcription mechanism(s) that lead to cell division.Specifications
| CDK8 | |
| Polyclonal | |
| Unconjugated | |
| Cdk8 | |
| Cdk8; CDK8 protein kinase; Cell division protein kinase 8; cyclin dependent kinase 8; Cyclin-dependent kinase 8; K35; Mediator complex subunit CDK8; Mediator of RNA polymerase II transcription subunit CDK8; MGC126074; MGC126075; Protein kinase K35; RGD1560888; wu:fk95h11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1024, 264064 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot | |
| Lot-Specific | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| P49336, Q8R3L8 | |
| Cdk8 | |
| This antibody was raised against a peptide within the C-terminal region of human CDk8. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction