Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDNF Antibody (104), PE, Novus Biologicals™
SDP

Catalog No. NB333657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333657 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333657 Supplier Novus Biologicals Supplier No. NBP290322PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CDNF Monoclonal antibody specifically detects CDNF in Human samples. It is validated for ELISA

Specifications

Antigen CDNF
Applications ELISA
Classification Monoclonal
Clone 104
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias arginine-rich, mutated in early stage tumors-like 1, ARMETL1, ARMET-like protein 1, cerebral dopamine neurotrophic factor, Conserved dopamine neurotrophic factor
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human CDNF (Accession#: NP_001025125.2, Met1-Leu187)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 441549
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.