Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDT1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB1002567C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002567C 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002567C Supplier Novus Biologicals Supplier No. NB1002567C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDT1 Polyclonal specifically detects CDT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen CDT1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chromatin licensing and DNA replication factor 1, DNA replication factor Cdt1, Double parked homolog, Double parked, Drosophila, homolog of, DUPRIS2
Gene Symbols CDT1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 500 and the C-terminus (residue 546) of human Chromatin Licensing and DNA Replication Factor 1 using the numbering given in entry NP_112190.1 (GeneID 81620).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81620
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.