Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDw17 Antibody (HO18.3G-6.F5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2452790.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CDw17 Monoclonal antibody specifically detects CDw17 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
CDw17 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
ST3GAL5 | |
B-2 Microglobulin associated proteins from a detergent lysate of human PBL | |
0.2 mg | |
Primary | |
CD17 is an intermediate glycosphingolipid from the metabolism of higher gangliosides that localizes to sphingolipid-sterol rafts. CD17 is detectable in monocytes, granulocytes, basophils, platelets, a subset of peripheral B cells (CD19+) and tonsil dendritic cells. It is rapidly down regulated on activated granulocytes and is upregulated on IL-2 activated T lymphocytes. CD17 binds to bacteria and may function in phagocytosis. VEGF-treated endothelial cells can produce CD17, which can then mediate signaling toward PECAM-1 expression and angiogenesis. Tumor necrosis factor alpha (TNFalpha)-induced astrogliosis (astrocyte proliferation and glial fibrillary acidic protein (GFAP) upregulation) in response to neuro-inflammation (i.e. spinal cord injury) causes an increase in intracellular levels of CD17. Aberrant levels of glycosphingolipids are a feature of cancer cells and may influence integrin clustering and internalization. | |
Store at 4C. | |
IgM |
Flow Cytometry, Immunofluorescence | |
HO18.3G-6.F5 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
8869 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction