Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDw60 Antibody (UM4D4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2451900.1MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence
CDw60 Monoclonal specifically detects CDw60 in Human samples. It is validated for ELISA.Specifications
CDw60 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Mouse | |
Protein A or G purified | |
RUO | |
This MAb reacts with 9-O-acetylated disialosyl group linked to the glycoprotein CD60w. CDw60 is a carbohydrate antigen present on gangliosides, platelets and a subset of CD4+, CD8+peripheral T-cells, but not on B-cells, monocytes or granulocytes. CDw60 is involved in the co-stimulation of T-cells. | |
Store at 4C. | |
IgM |
Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence | |
UM4D4 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Human rheumatoid synovial T cell line (ST-1) | |
0.1 mg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction