Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDw60 Antibody (UM4D4), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $487.50
Specifications
Antigen | CDw60 |
---|---|
Clone | UM4D4 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24519000
![]() |
Novus Biologicals
NBP2451900.02MG |
0.02 mg |
Each for $243.50
|
|
|||||
NBP24519001
![]() |
Novus Biologicals
NBP2451900.1MG |
0.1 mg |
Each for $487.50
|
|
|||||
NBP24519002
![]() |
Novus Biologicals
NBP2451900.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence
CDw60 Monoclonal specifically detects CDw60 in Human samples. It is validated for ELISA.Specifications
CDw60 | |
0.2 mg/mL | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
Human rheumatoid synovial T cell line (ST-1) | |
Primary | |
Store at 4C. |
UM4D4 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
IgM | |
Protein A or G purified | |
This MAb reacts with 9-O-acetylated disialosyl group linked to the glycoprotein CD60w. CDw60 is a carbohydrate antigen present on gangliosides, platelets and a subset of CD4+, CD8+peripheral T-cells, but not on B-cells, monocytes or granulocytes. CDw60 is involved in the co-stimulation of T-cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title