Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdx1 Antibody (123a), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP20160A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20160A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20160A70 Supplier Novus Biologicals Supplier No. NBP250160AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdx1 Monoclonal specifically detects Cdx1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cdx1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 123a
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915
Gene Symbols CDX1
Host Species Mouse
Immunogen Human Cdx1 N-terminal peptide
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1044
Test Specificity Clone 123a has a degree of cross reactivity with a determinant of different molecular weight when applied by WB and IHC in fibroblasts and smooth muscle cells. The cross reacting product does not appear in epithelial cells (or at least is significantly weaker).
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.