Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdx1 Antibody (123a), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309825 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309825 Supplier Novus Biologicals Supplier No. NBP250160MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdx1 Monoclonal antibody specifically detects Cdx1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP), ChIP assay Sequencing

Specifications

Antigen Cdx1
Applications Western Blot, Immunocytochemistry, ChIP Assay, ChIP sequencing (ChIP-seq)
Classification Monoclonal
Clone 123a
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing
Formulation 50mM Sodium Borate
Gene Alias caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915
Host Species Mouse
Immunogen Human Cdx1 N-terminal peptide
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1044
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.