Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2 Antibody (CDX2/2951R), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB391066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391066 Supplier Novus Biologicals Supplier No. NBP308738FR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen CDX2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CDX2/2951R
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
Host Species Rabbit
Immunogen Recombinant fragment (around aa150-249) of human CDX2 protein (exact sequence is proprietary) (Uniprot: Q99626)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1045
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.