Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB1002136F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002136F 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002136F Supplier Novus Biologicals Supplier No. NB1002136F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDX2 Polyclonal specifically detects CDX2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen CDX2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
Gene Symbols CDX2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human Caudal Type Homeobox Transcription Factor 2 using the numbering given in entry NP_001256.1 (GeneID 1045).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1045
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.