Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDX4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$330.00 - $547.00
Specifications
Antigen | CDX4 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CDX4 Polyclonal antibody specifically detects CDX4 in Human samples. It is validated for ImmunofluorescenceSpecifications
CDX4 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
caudal type homeo box transcription factor 4, caudal type homeobox 4, caudal type homeobox transcription factor 4, Caudal-type homeobox protein 4, homeobox protein CDX-4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: GSPMPASNFAAAPAFSHYMGYPHMPSMDPHWPSLGVWGSPYSPPREDWSVYPGPSSTMGTVPVNDVTSSPAAFCSTDYSNLG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
1046 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title