Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDYL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152986
Description
CDYL Polyclonal specifically detects CDYL in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CDYL | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CDYL1bA620A17.2 (chromodomain protein, Y chromosome-like), CDY-like, CDY-like, autosomal, chromodomain protein, Y chromosome-like, chromodomain protein, Y-like, chromodomain Y-like protein, DKFZP586C1622, EC 2.3.1.48, MGC131936, testis-specific chromodomain Y-like protein | |
Rabbit | |
Affinity purified | |
RUO | |
9425 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence | |
Q9Y232 | |
CDYL | |
Synthetic peptides corresponding to CDYL(chromodomain protein, Y-like) The peptide sequence was selected from the n terminal of CDYL. Peptide sequence YIHDFNRRHTEKQKESTLTRTNRTSPNNARKQISRSTNSNFSKTSPKALV. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Canine: 100%; Equine: 100%; Human: 100%; Bovine: 92%; Chicken: 92%; Guinea pig: 92%; Mouse: 78%; Rat: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction