Learn More
Invitrogen™ CDYL Polyclonal Antibody

Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Specifications
Specifications
| Antigen | CDYL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | CDYL |
| Gene Accession No. | Q9Y232 |
| Gene Alias | AI325931; CDYL; CDYL1; CDY-like; CDY-like, autosomal; chromodomain protein Y-like variant 2; chromodomain protein Y-like variant 3; chromodomain protein Y-like variant 4; chromodomain protein, Y chromosome-like; chromodomain protein, Y-like; chromodomain Y like; chromodomain Y-like protein; chromodomain Y-like protein pseudogene; Crotonyl-CoA hydratase; DKFZp586C1622; MGC131936; Putative histone acetyltransferase Cdyl; Putative tubulin acetyltransferase Cdyl; testis-specific chromodomain Y-like protein |
| Gene Symbols | CDYL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.