Learn More
Description
Specifications
Specifications
| Antigen | CEA |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | PARLAM 4 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | Carcino Embryonic Antigen, Carcinoembryonic antigen |
| Host Species | Mouse |
| Immunogen | Clone PARLAM 4 is a mouse monoclonal IgG1 antibody derived by fusion of Sp 2/0 Ag 14 mouse myeloma cells with spleen cells from a BABL/c mouse immunized with isolated human CEA. The immunogen has been isolated from human colonic carcinoma cells. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
