Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEA Antibody (PARLAM 4), PE, Novus Biologicals™
SDP

Catalog No. NBP197718PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197718PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197718PE Supplier Novus Biologicals Supplier No. NBP197718PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEA Monoclonal specifically detects CEA in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CEA
Applications Flow Cytometry
Classification Monoclonal
Clone PARLAM 4
Conjugate PE
Dilution Flow Cytometry
Gene Alias Carcino Embryonic Antigen, Carcinoembryonic antigen
Host Species Mouse
Immunogen Clone PARLAM 4 is a mouse monoclonal IgG1 antibody derived by fusion of Sp 2/0 Ag 14 mouse myeloma cells with spleen cells from a BABL/c mouse immunized with isolated human CEA. The immunogen has been isolated from human colonic carcinoma cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Most polyclonal CEA antisera show cross-reactivity with related antigens such as biliary glycoprotein (BGP) and non-specific cross-reacting antigen 1/11 (NCA). This antibody does not show cross reactivity, neither with BGP nor with NCA. In immunoblotting the antibody recognizes a single band of 180 kD.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.