Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CD66a Antibody (283340), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB349148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349148 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349148 Supplier Novus Biologicals Supplier No. FAB2244MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen CEACAM1/CD66a
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 283340
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CEACAM-1, Gln35-Gly428, Accession # P13688
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 634
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.