Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CD66a Antibody (28T25), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP221997FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221997FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221997FR Supplier Novus Biologicals Supplier No. NBP221997FR

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen CEACAM1/CD66a
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 28T25
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Gene Symbols CEACAM1
Host Species Mouse
Immunogen Human recombinant CEACAM1/CD66a
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 634
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.