Learn More
Description
Specifications
Specifications
| Antigen | CEACAM1/CD66a |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | C5-1X |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 5 μg/mL, Flow Cytometry 10 μg/mL, ELISA 10 μg/mL, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Sandwich ELISA 1:100-1:2000, CyTOF-ready |
| Gene Alias | antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen |
| Gene Symbols | CEACAM1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
