Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CD66a Antibody (CC1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP143390R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143390R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143390R Supplier Novus Biologicals Supplier No. NBP143390R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CEACAM1/CD66a
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone CC1
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Gene Symbols CEACAM1
Host Species Mouse
Immunogen The immunogen for this antibody was CD66a.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 634
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.