Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CD66a Antibody (F7), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N259673PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N259673PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N259673PEC7 Supplier Novus Biologicals Supplier No. NBP259673PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CEACAM1/CD66a
Applications Flow Cytometry
Classification Monoclonal
Clone F7
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Gene Symbols CEACAM1
Host Species Mouse
Immunogen The immunogen for this antibody is proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 634.0
Test Specificity Binds specific to the N-domain of rat CEACAM1. Does not cross-react with human CEACAM-1, -3. -4, -5, -6, 7 and -8, mouse CEACAM1 and mouse CEACAM2.
Target Species Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.