Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CD66a Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP14719002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14719002 0.2 mg
NB018644 0.02 mg
NBP14719001 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14719002 Supplier Novus Biologicals Supplier No. NBP1471900.2MG

Rabbit Polyclonal Antibody

CEACAM1/CD66a Polyclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen CEACAM1/CD66a
Applications Western Blot, Flow Cytometry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml, Flow Cytometry 2-10 ug/ml, ELISA 1-5 ug/ml
Formulation PBS (pH 7.4) with No Preservative
Gene Accession No. P13688
Gene Alias antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Gene Symbols CEACAM1
Host Species Rabbit
Immunogen Recombinant human sCEA-1 (D1-4)
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 634
Test Specificity The polyclonal antibody will detect recombinant human soluble CEACAM-1 in WB and ELISA
Reconstitution Reconstitute in sterile water to a concentration of 0.1-1.0mg/mL.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.