Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEACAM1/CEACAM5 Recombinant Rabbit Monoclonal Antibody (BLR032F)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | CEACAM1/CEACAM5 |
---|---|
Clone | BLR032F |
Concentration | 50 μg/mL |
Content And Storage | 4°C |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544259
|
Invitrogen™
MA544259 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544260
|
Invitrogen™
MA544260 |
10 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
Over-expression of CEA/CEACAM5 in tumors of epithelial origin is the basis of its wide-spread use as a tumor marker CEACAM1 expression is down-regulated in many tumors indicating a tumor-suppressive function. The anti-tumor effect may be due to inhibition of tumor angiogenesis, possibly by increased secretion of anti-angiogenic molecules from the cells. The function of CEA family members varies widely: they function as cell adhesion molecules, tumor suppressors, regulators of lymphocyte and dendritic cell activation, receptors of Neisseria species and other bacteria.Specifications
CEACAM1/CEACAM5 | |
50 μg/mL | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
P06731, P13688 | |
1048, 634 | |
Recombinant protein representing the extracellular domain of human CEACAM1 (residues 35-433). | |
Antibody |
BLR032F | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
CEACAM1 | |
antigen CD66; BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); carcinoembryonic antigen-related cell adhesion molecule 5; CD66a; CD66a antigen; CD66e; CEA; CEACAM1; CEACAM5; Meconium antigen 100; pan CEACAM; sCD66e; soluble CD66e | |
CEACAM1, CEACAM5 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title