Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM16 Antibody (2747B), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB295299 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295299 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295299 Supplier Novus Biologicals Supplier No. FAB9885MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CEACAM16 Monoclonal specifically detects CEACAM16 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen CEACAM16
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2747B
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 16
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO-derived human CEACAM-16, Met1-Gly425, Accession # Q2WEN9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388551
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.