Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM18 Antibody (1057633), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB302280 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302280 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302280 Supplier Novus Biologicals Supplier No. FAB9869L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM18 Monoclonal antibody specifically detects CEACAM18 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CEACAM18
Applications Flow Cytometry
Classification Monoclonal
Clone 1057633
Conjugate DyLight 550
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Carcinoembryonic Antigen-Related Cell Adhesion Molecule 18
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human CEACAM-18, Gln31-His317, Accession # A8MTB9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729767
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.