Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM19 Antibody (1042108), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB302407 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302407 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302407 Supplier Novus Biologicals Supplier No. FAB93971E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM19 Monoclonal specifically detects CEACAM19 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CEACAM19
Applications Flow Cytometry
Classification Monoclonal
Clone 1042108
Conjugate DyLight 405
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 19
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CEACAM-19, Met1-Gly157, Accession # Q7Z692
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56971
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.