Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM19 Antibody (Z5P2D9*A9), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB253124AFD Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253124AFD 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB253124AFD Supplier Novus Biologicals Supplier No. NBP253124AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM19 Monoclonal specifically detects CEACAM19 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CEACAM19
Applications Western Blot, ELISA
Classification Monoclonal
Clone Z5P2D9*A9
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 19
Gene Symbols CEACAM19
Host Species Mouse
Immunogen Peptide sequence - RSINPARPLP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56971
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.