Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM19 Antibody (Z5P2D9*A9), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309131 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309131 Supplier Novus Biologicals Supplier No. NBP253124MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM19 Monoclonal antibody specifically detects CEACAM19 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen CEACAM19
Applications Western Blot, ELISA
Classification Monoclonal
Clone Z5P2D9*A9
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 19
Host Species Mouse
Immunogen Peptide sequence - RSINPARPLP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56971
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.