Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM2 Antibody (935610), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB310532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310532 Supplier Novus Biologicals Supplier No. FAB9214PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CEACAM2 Monoclonal antibody specifically detects CEACAM2 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen CEACAM2
Applications Flow Cytometry
Classification Monoclonal
Clone 935610
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias Bgp;Bgp2;Carcinoembryonic antigen-related cell adhesion molecule 2;Ceacam2-L
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2, Gln35-Asp422, Accession # Q925P2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26367
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.