Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM20 Antibody (1032339), HRP, Novus Biologicals™
SDP

Catalog No. NB345736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345736 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345736 Supplier Novus Biologicals Supplier No. FAB9416H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM20 Monoclonal specifically detects CEACAM20 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CEACAM20
Applications Flow Cytometry
Classification Monoclonal
Clone 1032339
Conjugate HRP
Dilution Flow Cytometry
Formulation PBS
Gene Alias UNQ9366
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human CEACAM-20, Gln31-Gly450, Accession # Q6UY09-1
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125931
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.