Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM21 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB329521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329521 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329521 Supplier Novus Biologicals Supplier No. NBP297874MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEACAM21 Polyclonal antibody specifically detects CEACAM21 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CEACAM21
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 21, CEACAM3, FLJ13540, MGC119874, R29124_1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CEACAM21 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 90273
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.