Learn More
Description
Specifications
Specifications
| Antigen | CEACAM3/CD66d |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD66D, CEA, CEACAM3 carcinoembryonic antigen-related cell adhesion molecule 3, CGM1, W264, W282 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant CEACAM3/CD66d (NP_001806.2; Met1-Gly155). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
