Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM4 Antibody (822608), HRP, Novus Biologicals™
SDP

Catalog No. NB346399 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346399 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346399 Supplier Novus Biologicals Supplier No. FAB7845H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM4 Monoclonal specifically detects CEACAM4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CEACAM4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 822608
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias carcinoembryonic antigen CGM7, carcinoembryonic antigen gene family member 7, carcinoembryonic antigen-related cell adhesion molecule 4, CGM7, CGM7_HUMAN
Host Species Mouse
Immunogen E. coli-derived recombinant human CEACAM-4, Phe36-Val140, Accession # O75871
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1089
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.