Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5/CD66e Antibody (061), Biotin, Novus Biologicals™
SDP

Catalog No. NB067943 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067943 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067943 Supplier Novus Biologicals Supplier No. NBP289785B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CEACAM5/CD66e
Applications Flow Cytometry
Classification Monoclonal
Clone 61
Conjugate Biotin
Dilution Flow Cytometry
Formulation PBS
Gene Alias Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM5/CD66e (NP_004354.2; Met1-Ala685).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1048
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.