Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5/CD66e Antibody (OTI3D8), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP270407UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270407UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270407UV Supplier Novus Biologicals Supplier No. NBP270407UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CEACAM5/CD66e
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3D8
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Gene Symbols CEACAM5
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 35-680 of human CEACAM5/CD66e (NP_004354) produced in SF9 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1048.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.