Learn More
Description
Specifications
Specifications
| Antigen | CEACAM5/CD66e |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SPM584 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Alias | Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 |
| Gene Symbols | CEACAM5 |
| Host Species | Mouse |
| Immunogen | Human colon carcinoma extract (Uniprot: P06731) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
