Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM6/CD66c Antibody (408), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB099622 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB099622 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB099622 Supplier Novus Biologicals Supplier No. NBP289669AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CEACAM6/CD66c Monoclonal specifically detects CEACAM6/CD66c in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CEACAM6/CD66c
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 408
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM Sodium Borate
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM6/CD66c (NP_002474.3; Met 1-Gly 320).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4680
Test Specificity No cross-reactivity in ELISA with: Human CEACAM1/ CD66a Human CEACAM5/ CD66e
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.