Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM6/CD66c Antibody (439424), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB312662 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312662 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312662 Supplier Novus Biologicals Supplier No. FAB3934PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM6/CD66c Monoclonal specifically detects CEACAM6/CD66c in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CEACAM6/CD66c
Applications Flow Cytometry
Classification Monoclonal
Clone 439424
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CEACAM-6/CD66c, Lys35-Gly320, Accession # P40199
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4680
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.