Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CEACAM6/CD66c Antibody Pair [Biotin]
SDP

Catalog No. NB009645 Shop All R&D Systems Products
Click to view available options
Quantity:
15 Plates

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human CEACAM6/CD66c.

Specifications

Assay Range 15.6 pg/mL
Assay Sensitivity 15.6-1000 pg/mL
Assay Sandwich ELISA
Conjugate Biotin
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human CEACAM6/CD66c.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Gene ID (Entrez) 4680
Gene Symbol CEACAM6
Host Species Rabbit/Mouse
Kit Contents
  • Detection Antibody: 0.4 mg/mL of biotinylated rabbit anti-CEACAM6/CD66c polyclonal antibody.
  • Capture Antibody: 1 mg/mL of mouse anti-CEACAM6/CD66c monoclonal antibody.
  • Standard: Each vial contains 88 ng of recombinant CEACAM6/CD66c.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target CEACAM6/CD66c
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.