Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM6/CD66c Rabbit anti-Human, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289671100UL
Description
CEACAM6/CD66c Monoclonal specifically detects CEACAM6/CD66c in Human samples. It is validated for ELISA.Specifications
CEACAM6/CD66c | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
2 | |
ELISA 1:5000-1:10000 | |
carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen | |
Recombinant Human CEACAM6/CD66c Protein | |
100 μg | |
Cancer | |
4680 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction